Stampa…

Anti-CGB5 Mouse Monoclonal Antibody

Fornitore: Biorbyt
BIRBORB27487-1EA 0 EUR
BIRBORB27487-1
Anti-CGB5 Mouse Monoclonal Antibody
Anticorpi
Type: Primary
Antigen: CGB5 (chorionic gonadotropin, beta polypeptide 5)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR