Stampa…

Anti-Neurofilaments 200 kDa Mouse Monoclonal Antibody

Fornitore: Biorbyt
BIRBORB27437-1EA 0 EUR
BIRBORB27437-1
Anti-Neurofilaments 200 kDa Mouse Monoclonal Antibody
Anticorpi
Type: Primary
Antigen: Neurofilaments 200 kDa
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity:
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR