Stampa…

Anti-AGER Rabbit Polyclonal Antibody

Fornitore: Biorbyt
BIRBORB27389-100EA 0 EUR
BIRBORB27389-100
Anti-AGER Rabbit Polyclonal Antibody
Anticorpi
Immunochemistry: 1-5 ?g/ml; Western Blot: 1-2 ?g/ml

Type: Primary
Antigen: AGER (advanced glycosylation end product-specific receptor)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR