Stampa…

Anti-AGER Goat Polyclonal Antibody

Fornitore: Biorbyt
BIRBORB27365-100EA 0 EUR
BIRBORB27365-100
Anti-AGER Goat Polyclonal Antibody
Anticorpi
Immunochemistry: (P) (1:400); ELISA: (1:10000-1:260000); Western Blot: 1:2000

Type: Primary
Antigen: AGER (advanced glycosylation end product-specific receptor)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity:
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR