Stampa…

Anti-COL2 Rabbit Polyclonal Antibody

Fornitore: Biorbyt
BIRBORB27314-0.5EA 0 EUR
BIRBORB27314-0.5
Anti-COL2 Rabbit Polyclonal Antibody
Anticorpi
ImmunofluorescenceA 1:40; Immunochemistry: 1:100 - 1:500

Type: Primary
Antigen: COL2 (collagen, type II)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR