Stampa…

Anti-Cytokeratin (High Molecular Weight) 34betaE12 Mouse Monoclonal Antibody

Fornitore: Biorbyt
BIRBORB27195-1EA 0 EUR
BIRBORB27195-1
Anti-Cytokeratin (High Molecular Weight) 34betaE12 Mouse Monoclonal Antibody
Anticorpi
Type: Primary
Antigen: Cytokeratin (High Molecular Weight) 34betaE12
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR