Stampa…

Anti-COL4 Goat Polyclonal Antibody

Fornitore: Biorbyt
BIRBORB26834-0.2EA 0 EUR
BIRBORB26834-0.2
Anti-COL4 Goat Polyclonal Antibody
Anticorpi
ELISA: 1:10-1:20

Type: Primary
Antigen: COL4 (collagen, type IV)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity:
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR