Stampa…

Anti-Nidogen 2 Rabbit Polyclonal Antibody

Fornitore: Biorbyt
BIRBORB26826-20EA 0 EUR
BIRBORB26826-20
Anti-Nidogen 2 Rabbit Polyclonal Antibody
Anticorpi
Type: Primary
Antigen: Nidogen 2
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Mouse
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR