Stampa…

Anti-Staph. Enterotoxin E Rabbit Polyclonal Antibody

Fornitore: Biorbyt
BIRBORB26797-1EA 0 EUR
BIRBORB26797-1
Anti-Staph. Enterotoxin E Rabbit Polyclonal Antibody
Anticorpi
Type: Primary
Antigen: Staph. Enterotoxin E
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR