Stampa…

Anti-D5 Dopamine Rec. Rabbit Polyclonal Antibody

Fornitore: Biorbyt
BIRBORB26727-100EA 0 EUR
BIRBORB26727-100
Anti-D5 Dopamine Rec. Rabbit Polyclonal Antibody
Anticorpi
Immunochemistry: 1:2500; Western Blot: 1:200

Type: Primary
Antigen: D5 Dopamine Rec.
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR