Stampa…
Anti-D2L Dopamine Receptor Rabbit Polyclonal Antibody
Fornitore: Biorbyt
BIRBORB26714-100EA 0 EUR
BIRBORB26714-100
Anti-D2L Dopamine Receptor Rabbit Polyclonal Antibody
Anticorpi
Western Blot: 1:400
Type: Primary
Antigen: D2L Dopamine Receptor
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Visualizza i codici