Stampa…
Anti-M3 Muscarinic Rec. Rabbit Polyclonal Antibody
Fornitore: Biorbyt
BIRBORB26703-100EA 0 EUR
BIRBORB26703-100
Anti-M3 Muscarinic Rec. Rabbit Polyclonal Antibody
Anticorpi
Immunochemistry: 1:2500; Western Blot: 1:500
Type: Primary
Antigen: M3 Muscarinic Rec.
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Visualizza i codici