Stampa…

Anti-CD41 gpIIb IIIa Mouse Monoclonal Antibody (FITC (Fluorescein))

Fornitore: Biorbyt
BIRBORB26619-100EA 0 EUR
BIRBORB26619-100
Anti-CD41 gpIIb IIIa Mouse Monoclonal Antibody (FITC (Fluorescein))
Anticorpi
Immunochemistry: 1:50-1:100

Type: Primary
Antigen: CD41 gpIIb IIIa
Clonality: monoclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR