Stampa…
Anti-Mouse VCAM-1 Rat Monoclonal Antibody
Fornitore: Biorbyt
BIRBORB26551-0.5EA 0 EUR
BIRBORB26551-0.5
Anti-Mouse VCAM-1 Rat Monoclonal Antibody
Anticorpi
Type: Primary
Antigen: Mouse VCAM-1
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG1 Kappa (Rat)
Reactivity: Mouse