Stampa…

Anti-Rat CD11b Mouse Monoclonal Antibody

Fornitore: Biorbyt
BIRBORB26550-0.5EA 0 EUR
BIRBORB26550-0.5
Anti-Rat CD11b Mouse Monoclonal Antibody
Anticorpi
Type: Primary
Antigen: Rat CD11b
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Rat
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR