Stampa…

Anti-Mouse CD71 Rat Monoclonal Antibody (FITC (Fluorescein))

Fornitore: Biorbyt
BIRBORB26430-0.5EA 0 EUR
BIRBORB26430-0.5
Anti-Mouse CD71 Rat Monoclonal Antibody (FITC (Fluorescein))
Anticorpi
Type: Primary
Antigen: Mouse CD71
Clonality: monoclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rat
Isotype: IgG2a (Rat)
Reactivity: Mouse
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR