Stampa…

Anti-Mouse CD18 (LFA-1 Beta) Rat Monoclonal Antibody

Fornitore: Biorbyt
BIRBORB26386-0.5EA 0 EUR
BIRBORB26386-0.5
Anti-Mouse CD18 (LFA-1 Beta) Rat Monoclonal Antibody
Anticorpi
Flow Cytometry: 1 ?g

Type: Primary
Antigen: Mouse CD18 (LFA-1 Beta)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG2a (Rat)
Reactivity: Mouse
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR