Stampa…

Anti-ITGAX Mouse Monoclonal Antibody (Biotin)

Fornitore: Biorbyt
BIRBORB26354-0.2EA 0 EUR
BIRBORB26354-0.2
Anti-ITGAX Mouse Monoclonal Antibody (Biotin)
Anticorpi
Type: Primary
Antigen: ITGAX (integrin, alpha X (complement component 3 receptor 4 subunit))
Clonality: monoclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR