Stampa…

Anti-F12 Mouse monoclonal antibody

Fornitore: Biorbyt
BIRBORB26282-0.25EA 0 EUR
BIRBORB26282-0.25
Anti-F12 Mouse monoclonal antibody
Anticorpi
Type: Primary
Antigen: F12 (coagulation factor XII (Hageman factor))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR