Stampa…

Anti-SETBP1 Mouse Monoclonal Antibody

Fornitore: Biorbyt
BIRBORB26182-1EA 0 EUR
BIRBORB26182-1
Anti-SETBP1 Mouse Monoclonal Antibody
Anticorpi
Type: Primary
Antigen: SETBP1 (SET binding protein 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR