Stampa…

Anti-CD59 Mouse Monoclonal Antibody (Biotin)

Fornitore: Biorbyt
BIRBORB26126-100EA 0 EUR
BIRBORB26126-100
Anti-CD59 Mouse Monoclonal Antibody (Biotin)
Anticorpi
Flow Cytometry: 1:40

Type: Primary
Antigen: CD59 (CD59 molecule, complement regulatory protein)
Clonality: monoclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR