Stampa…

Anti-CRP Mouse Monoclonal Antibody (Biotin)

Fornitore: Biorbyt
BIRBORB26031-1EA 0 EUR
BIRBORB26031-1
Anti-CRP Mouse Monoclonal Antibody (Biotin)
Anticorpi
Type: Primary
Antigen: CRP (C-reactive protein, pentraxin-related)
Clonality: monoclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR