Stampa…

Anti-FOLH1 Mouse monoclonal antibody

Fornitore: Biorbyt
BIRBORB25921-0.5EA 0 EUR
BIRBORB25921-0.5
Anti-FOLH1 Mouse monoclonal antibody
Anticorpi
Type: Primary
Antigen: FOLH1 (folate hydrolase (prostate-specific membrane antigen) 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity:
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR