Stampa…

Anti-CEACAM5 Mouse monoclonal antibody

Fornitore: Biorbyt
BIRBORB25861-1EA 0 EUR
BIRBORB25861-1
Anti-CEACAM5 Mouse monoclonal antibody
Anticorpi
Type: Primary
Antigen: CEACAM5 (carcinoembryonic antigen-related cell adhesion molecule 5)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR