Stampa…

Anti-Malondialdehyde (MDA) Goat Polyclonal Antibody

Fornitore: Biorbyt
BIRBORB25780-1EA 0 EUR
BIRBORB25780-1
Anti-Malondialdehyde (MDA) Goat Polyclonal Antibody
Anticorpi
Western Blot: 1:10000-1:160000; ELISA: 1:10000-1:160000

Type: Primary
Antigen: Malondialdehyde (MDA)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity:
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR