Stampa…

Anti-Factor H (Beta 1H) Sheep Polyclonal Antibody

Fornitore: Biorbyt
BIRBORB25711-1EA 0 EUR
BIRBORB25711-1
Anti-Factor H (Beta 1H) Sheep Polyclonal Antibody
Anticorpi
Type: Primary
Antigen: Factor H (Beta 1H)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype: none
Reactivity:
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR