Stampa…

Anti-T-cadherin Rabbit Polyclonal Antibody

Fornitore: Biorbyt
BIRBORB25623-100EA 0 EUR
BIRBORB25623-100
Anti-T-cadherin Rabbit Polyclonal Antibody
Anticorpi
Immunoprecipitation: 20 ?g/ml; Western Blot: 0.5-1 ?g/ml

Type: Primary
Antigen: T-cadherin
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR