Stampa…

Anti-APOC2 Goat Polyclonal Antibody

Fornitore: Biorbyt
BIRBORB25556-1EA 0 EUR
BIRBORB25556-1
Anti-APOC2 Goat Polyclonal Antibody
Anticorpi
ELISA: 1:200-1:80000

Type: Primary
Antigen: APOC2 (apolipoprotein C-II)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity:
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR