Stampa…

Anti-APOC1 Rabbit Polyclonal Antibody

Fornitore: Biorbyt
BIRBORB25548-0.5EA 0 EUR
BIRBORB25548-0.5
Anti-APOC1 Rabbit Polyclonal Antibody
Anticorpi
DB: 1:200-1:5000; ELISA: 1:5000-1:20000

Type: Primary
Antigen: APOC1 (apolipoprotein C-I)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR