Stampa…

Anti-Glucose Oxidase Rabbit Polyclonal Antibody

Fornitore: Biorbyt
BIRBORB25360-50EA 0 EUR
BIRBORB25360-50
Anti-Glucose Oxidase Rabbit Polyclonal Antibody
Anticorpi
ELISA: 1:5000-1:30000

Type: Primary
Antigen: Glucose Oxidase
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR