Stampa…

Anti-AMY2B Rabbit Polyclonal Antibody

Fornitore: Biorbyt
BIRBORB25304-1EA 0 EUR
BIRBORB25304-1
Anti-AMY2B Rabbit Polyclonal Antibody
Anticorpi
Western Blot: 1:8000

Type: Primary
Antigen: AMY2B (amylase, alpha 2B (pancreatic))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR