Stampa…

Anti-APOA1 Goat Polyclonal Antibody (Biotin)

Fornitore: Biorbyt
BIRBORB25280-1EA 0 EUR
BIRBORB25280-1
Anti-APOA1 Goat Polyclonal Antibody (Biotin)
Anticorpi
ELISAl: 1:200-1:20000; DB: 1:100-1:4000

Type: Primary
Antigen: APOA1 (apolipoprotein A-I)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: none
Reactivity:
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR