Stampa…

Anti-Hamster Apo AI Rabbit Polyclonal Antibody

Fornitore: Biorbyt
BIRBORB25177-1EA 0 EUR
BIRBORB25177-1
Anti-Hamster Apo AI Rabbit Polyclonal Antibody
Anticorpi
Type: Primary
Antigen: Hamster Apo AI
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Hamster
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR