Stampa…

Anti-APOE Rabbit Polyclonal Antibody

Fornitore: Biorbyt
BIRBORB25175-1EA 0 EUR
BIRBORB25175-1
Anti-APOE Rabbit Polyclonal Antibody
Anticorpi
Western Blot: 1:500-1:1000

Type: Primary
Antigen: APOE (apolipoprotein E)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Mouse
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR