Stampa…

Anti-Lambda Light Chain Goat Polyclonal Antibody

Fornitore: Biorbyt
BIRBORB25148-10EA 0 EUR
BIRBORB25148-10
Anti-Lambda Light Chain Goat Polyclonal Antibody
Anticorpi
Type: Primary
Antigen: Lambda Light Chain
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR