Stampa…

Anti-APOC3 Goat Polyclonal Antibody

Fornitore: Biorbyt
BIRBORB25090-1EA 0 EUR
BIRBORB25090-1
Anti-APOC3 Goat Polyclonal Antibody
Anticorpi
Type: Primary
Antigen: APOC3 (apolipoprotein C-III)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR