Stampa…

Anti-MPO Mouse Monoclonal Antibody (FITC (Fluorescein))

Fornitore: Biorbyt
BIRBORB25000-100EA 0 EUR
BIRBORB25000-100
Anti-MPO Mouse Monoclonal Antibody (FITC (Fluorescein))
Anticorpi
Flow Cytometry: 1:10

Type: Primary
Antigen: MPO (myeloperoxidase)
Clonality: monoclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR