Stampa…

Anti-LEP Mouse Monoclonal Antibody

Fornitore: Biorbyt
BIRBORB24757-100EA 0 EUR
BIRBORB24757-100
Anti-LEP Mouse Monoclonal Antibody
Anticorpi
  • Immunofluorescence: 1:2000

Type: Primary
Antigen: LEP (leptin)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR