Stampa…

Anti-Filamentous Phages M13/fd/F1 Mouse Monoclonal Antibody (Biotin)

Fornitore: Biorbyt
BIRBORB24723-0.25EA 0 EUR
BIRBORB24723-0.25
Anti-Filamentous Phages M13/fd/F1 Mouse Monoclonal Antibody (Biotin)
Anticorpi
ELISA: 1:5000-10000

Type: Primary
Antigen: Filamentous Phages M13/fd/F1
Clonality: monoclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR