Stampa…

Anti-Penicillin + Derivatives Mouse Monoclonal Antibody

Fornitore: Biorbyt
BIRBORB24480-1EA 0 EUR
BIRBORB24480-1
Anti-Penicillin + Derivatives Mouse Monoclonal Antibody
Anticorpi
ELISA: 1:1500

Type: Primary
Antigen: Penicillin + Derivatives
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity:
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR