Stampa…

Anti-AVP Mouse Monoclonal Antibody

Fornitore: Biorbyt
BIRBORB24312-100EA 0 EUR
BIRBORB24312-100
Anti-AVP Mouse Monoclonal Antibody
Anticorpi
ELISA: 2 ?g/ml

Type: Primary
Antigen: AVP (arginine vasopressin)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR