Stampa…

Anti-Procalcitonin Mouse Monoclonal Antibody

Fornitore: Biorbyt
BIRBORB24289-1EA 0 EUR
BIRBORB24289-1
Anti-Procalcitonin Mouse Monoclonal Antibody
Anticorpi
Type: Primary
Antigen: Procalcitonin
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR