Stampa…

Anti-AGT Mouse Monoclonal Antibody

Fornitore: Biorbyt
BIRBORB24278-0.2EA 0 EUR
BIRBORB24278-0.2
Anti-AGT Mouse Monoclonal Antibody
Anticorpi
ELISA: 1:10-1:100

Type: Primary
Antigen: AGT (angiotensinogen (serpin peptidase inhibitor, clade A, member 8))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR