Stampa…

Anti-TSHB Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))

Fornitore: Biorbyt
BIRBORB24218-1EA 0 EUR
BIRBORB24218-1
Anti-TSHB Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Anticorpi
ELISA: 1:1000

Type: Primary
Antigen: TSHB (thyroid stimulating hormone, beta)
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Goat
Isotype: none
Reactivity:
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR