Stampa…

Anti-Thymosin beta-4 Rabbit Polyclonal Antibody

Fornitore: Biorbyt
BIRBORB24194-100EA 0 EUR
BIRBORB24194-100
Anti-Thymosin beta-4 Rabbit Polyclonal Antibody
Anticorpi
ELISA: 1:4000

Type: Primary
Antigen: Thymosin beta-4
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR