Stampa…

Anti-HBcAg (ayw) Mouse monoclonal antibody

Fornitore: Biorbyt
BIRBORB24148-100EA 0 EUR
BIRBORB24148-100
Anti-HBcAg (ayw) Mouse monoclonal antibody
Anticorpi
Type: Primary
Antigen: HBcAg (ayw)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR