Stampa…

Anti-Brucella abortus intact cells Mouse monoclonal antibody

Fornitore: Biorbyt
BIRBORB23967-1EA 0 EUR
BIRBORB23967-1
Anti-Brucella abortus intact cells Mouse monoclonal antibody
Anticorpi
Type: Primary
Antigen: Brucella abortus intact cells
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR