Stampa…

Anti-RSV, Long Strain Mouse Monoclonal Antibody

Fornitore: Biorbyt
BIRBORB23936-1EA 0 EUR
BIRBORB23936-1
Anti-RSV, Long Strain Mouse Monoclonal Antibody
Anticorpi
Type: Primary
Antigen: RSV, Long Strain
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR