Stampa…

Anti-Neisseria gonorrhoeae Mouse Monoclonal Antibody

Fornitore: Biorbyt
BIRBORB23867-100EA 0 EUR
BIRBORB23867-100
Anti-Neisseria gonorrhoeae Mouse Monoclonal Antibody
Anticorpi
Type: Primary
Antigen: Neisseria gonorrhoeae
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR