Stampa…

Anti-Toxoplasma gondii Mouse Monoclonal Antibody (FITC (Fluorescein))

Fornitore: Biorbyt
BIRBORB23795-100EA 0 EUR
BIRBORB23795-100
Anti-Toxoplasma gondii Mouse Monoclonal Antibody (FITC (Fluorescein))
Anticorpi
ELISA: 1:5-1:20

Type: Primary
Antigen: Toxoplasma gondii
Clonality: monoclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG2
Reactivity:
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR